missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl (1S,3S,5S)-3-Carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
tert-Butyl (1S,3S,5S)-3-Carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate
Specifications
Specifications
| Chemical Name or Material | tert-Butyl (1S,3S,5S)-3-Carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate |
| CAS | 361440-67-7 |
| Analyte or Component Names | tert-Butyl (1S,3S,5S)-3-Carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H18 N2 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 3-(aminocarbonyl)-, 1,1-dimethylethyl ester, (1S,3S,5S)- |
| InChI Formula | InChI=1S/C11H18N2O3/c1-11(2,3)16-10(15)13-7-4-6(7)5-8(13)9(12)14/h6-8H,4-5H2,1-3H3,(H2,12,14)/t6-,7-,8-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N1[C@H]2C[C@H]2C[C@H]1C(=O)N |
| IUPAC Name | tert-butyl (1S,3S,5S)-3-carbamoyl-2-azabicyclo[3.1.0]hexane-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction