missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 1-indolecarboxylate, 97%, Acros Organics™

Product Code. 15317390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15317390 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15317390 Supplier Thermo Scientific Acros Supplier No. 449790250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75400-67-8
Density 1.0700g/mL
Boiling Point 201.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Molecular Formula C13H15NO2
MDL Number MFCD02093939
Quantity 25 mL
Specific Gravity 1.07
Synonym 1-boc-indole, tert-butyl 1h-indole-1-carboxylate, n-boc-indole, tert-butyl 1-indolecarboxylate, 1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl indole, n-boc indole, pubchem7228, acmc-1bjgd, 1-tert-butoxycarbonylindole
InChI Key OWPIFQXNMLDXKW-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12
IUPAC Name tert-butyl 1H-indole-1-carboxylate
Molecular Weight (g/mol) 217.27
PubChem CID 3532980
Formula Weight 217.26
Percent Purity ≥96.0% (GC)
Chemical Name or Material tert-Butyl 1-indolecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.