missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%

Product Code. 11388166
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388166 1 g 1g
11398166 5 g 5g
11308176 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388166 Supplier Thermo Scientific Alfa Aesar Supplier No. L15144.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35000-38-5 | C24H25O2P | 376.44 g/mol

(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(tert-Butoxycarbonylmethylene)triphenylphosphorane is a chemical used in the synthesis of aldose reductase inhibitors, a podophyllotoxin derivative and tautomycin, a podophyllotoxin derivative and tautomycin. It Is also used in medicinal chemistry.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35000-38-5
Molecular Formula C24H25O2P
Molecular Weight (g/mol) 376.44
MDL Number MFCD00075545
InChI Key ZWZUFQPXYVYAFO-UHFFFAOYSA-N
Synonym tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
PubChem CID 256127
IUPAC Name tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 146°C to 150°C
Linear Formula (CH3)3CO2CCH=P(C6H5)3
Quantity 1 g
UN Number UN2811
Beilstein 2759875
Solubility Information Sparingly soluble in water.
Formula Weight 376.44
Percent Purity 98%
Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.