missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butoxycarbonylmethylene)triphenylphosphorane, 97%

Product Code. 10324412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10324412 1 g Glass bottle 1g
10038921 5 g Glass bottle 5g
10048921 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10324412 Supplier Thermo Scientific Supplier No. 370620010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35000-38-5 | C24H25O2P | 376.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35000-38-5
Molecular Formula C24H25O2P
Molecular Weight (g/mol) 376.44
MDL Number MFCD00075545
InChI Key ZWZUFQPXYVYAFO-UHFFFAOYSA-N
Synonym tert-butyl 2-triphenylphosphoranylidene acetate, tert-butoxycarbonylmethylene triphenylphosphorane, tert-butyl triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, 1,1-dimethylethyl ester, t-butoxycarbonylmethylene triphenylphosphorane, tert-butyl 2-triphenyl-??-phosphanylidene acetate, triphenylthoranylidene acetic acid 1,1-dimethylethyl ester, pubchem14371, tert-butyl 2-triphenyl, tert-butyl 2-triphenylylidene acetate
PubChem CID 256127
IUPAC Name tert-butyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES CC(C)(C)OC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 146.0°C to 151.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCO2C(CH3)3
Quantity 1 g
Formula Weight 376.44
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (tert-Butoxycarbonylmethylene)triphenylphosphorane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.