missing translation for 'onlineSavingsMsg'
Learn More

tert-Butoxybis(dimethylamino)methane, tech. 90%

Product Code. 11377469
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377469 25 g 25g
11357469 1 g 1g
11367469 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11377469 Supplier Thermo Scientific Alfa Aesar Supplier No. H27861.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5815-08-7 | C9H24N2O | 176.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5815-08-7
Molecular Formula C9H24N2O
Molecular Weight (g/mol) 176.30
MDL Number MFCD00042858
InChI Key HXRAMSFGUAOAJR-UHFFFAOYSA-P
Synonym tert-butoxy bis dimethylamino methane, 1-tert-butoxy-n,n,n',n'-tetramethylmethanediamine, tert-butoxybis dimethylamino methane, bredereck's reagent, bredereck reagent, tert-butoxy dimethylamino methyl dimethylamine, unii-jej0u558f7, bis dimethylamino-t-butoxymethane, tert-butoxy-bis dimethylamino methane, bis dimethylamino tert-butoxy methane
PubChem CID 79885
IUPAC Name N,N,N',N'-tetramethyl-1-[(2-methylpropan-2-yl)oxy]methanediamine
SMILES CC(C)(C)OC(N(C)C)N(C)C

Specifications

Density 0.844
Boiling Point 50°C to 55°C (15 mmHg)
Flash Point 41°C (106°F)
Linear Formula (CH3)3COCH[N(CH3)2]2
Refractive Index 1.422
Quantity 25 g
UN Number UN1993
Formula Weight 174.3
Percent Purity 90%
Grade Technical
Chemical Name or Material tert-Butoxybis(dimethylamino)methane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.