missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butanol, puriss. p.a. ACS Reagent, ≥99.7% (GC), Honeywell Riedel-de Haën™

Product Code. 15611710
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15611710 2.5 L Poly Bottle 2.5 L
15621710 500 mL Poly Bottle 500mL
15601710 1 L Poly Bottle 1 L
15631710 5 L Poly Bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15611710 Supplier Honeywell-Riedel-de Haen Supplier No. 194602.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS reagent,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25°C
Color Colorless
Density 0.775g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Packaging Poly Bottle
Physical Form Liquid
Refractive Index n20/D 1.387(lit.)
Linear Formula (CH3)3COH
Vapor Pressure 44mmHg (26°C), 31mmHg (20°C)
UN Number UN1120
Beilstein 906698
Boiling Point 83°C
Formula Weight 74.12g/mol
Percent Purity ≥99.7% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Tert-Butanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.