missing translation for 'onlineSavingsMsg'
Learn More

TERT Antibody (2C4) - Azide and BSA Free, Novus Biologicals™

Product Code. 18322970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322970 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322970 Supplier Novus Biologicals Supplier No. NBP280975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TERT Monoclonal specifically detects TERT in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TERT
Applications Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2C4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:500, Chromatin Immunoprecipitation reported in scientific literature (PMID 30936423), Flow Cytometry 1:50 - 1:200, Immunohistochemistry : 1:50, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26725521), Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin : 1:50, Chromatin Immunoprecipitation (ChIP)
Formulation Tris-Glycine
Gene Alias EST2Telomerase-associated protein 2, hEST2, hTERT, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7
Gene Symbols TERT
Host Species Mouse
Immunogen Full-length recombinant human Telomerase reverse transcriptase (TERT) from insect cells. [Uniprot: O14746].
Purification Method IgM purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7015
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.