missing translation for 'onlineSavingsMsg'
Learn More

tert-Amylamine, 98%, ACROS Organics™

Product Code. 10082130 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10082130

Brand: Thermo Scientific Acros 175611000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 594-39-8
Color Brown to Yellow
Density 0.7460g/mL
Boiling Point 77.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H13N
Linear Formula C2H5C(CH3)2NH2
Refractive Index 1.3986 to 1.4006
MDL Number MFCD00008056
Quantity 100 mL
Specific Gravity 0.746
Synonym tert-amylamine, tert-pentylamine, 2-butanamine, 2-methyl, t-amylamine, 1,1-dimethylpropylamine, 2-methyl-2-butylamine, 2-amino-2-methylbutane, 2-methyl-butan-2-amine, 1,1-dimethyl-1-propylamine, t-pentylamine
InChI Key GELMWIVBBPAMIO-UHFFFAOYSA-N
SMILES CCC(C)(C)N
IUPAC Name 2-methylbutan-2-amine
Molecular Weight (g/mol) 87.166
PubChem CID 68986
ChEBI CHEBI:84236
Formula Weight 87.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Amylamine, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.