missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)

Product Code. 10173243
Click to view available options
Quantity:
10,000 g
100 g
2000 g
500 g
Packaging Type:
Poly Bottle
Poly Pail
Unit Size:
100g
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 10173243

Brand: Fisher BioReagents™ BP2468100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.

Formulation Usage: 50.8g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Powder
Grade Molecular genetics
Media Type Terrific Broth
Product Type Microbiology Media
For Use With (Application) Cultivation of recombinant strains of Escherichia coli
Packaging Type Poly Bottle
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.