missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Terrific Broth, Modified

Product Code. 12811670
Change view
Click to view available options
Quantity:
600 g
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12811670 600 g 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12811670 Supplier Fisher BioReagents™ Supplier No. 12811670
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Buffered enriched medium used to obtain high yields of plasmid DNA from transformed E.coli.

  • Each capsule contains 24g of broth
  • Sterilize by autoclaving

Specifications

Broth Type Terrific Broth
Format Capsules
Product Type Microbiology Media
Packaging Type 25 Capsules
Quantity 600 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.