missing translation for 'onlineSavingsMsg'
Learn More

Terpineol (mixture of isomers) 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317942
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317942 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317942 Supplier Dr. Ehrenstorfer Supplier No. DREA17322340ME100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terpineol
CAS 8000-41-7
Analyte or Component Names Terpineol
Shipping Condition Room Temperature
Molecular Formula C10H18O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Alpha Jax, Terpineol 318, Terpineol C, Terpineol LW;
InChI Formula InChI=1S/3C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11;2*1-8(2)9-4-6-10(3,11)7-5-9/h4,9,11H,5-7H2,1-3H3;11H,4-7H2,1-3H3;9,11H,1,4-7H2,2-3H3
SMILES CC1=CCC(C(O)(C)C)CC1.CC2(O)CCC(C(C)=C)CC2.CC(CC/3)(O)CCC3=C(C)\C
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)cyclohexan-1-ol--1-methyl-4-(propan-2-ylidene)cyclohexan-1-ol--2-(4-methylcyclohex-3-en-1-yl)propan-2-ol (1/1/1)
Solvent or Matrix Methanol
Molecular Weight (g/mol) 154.25
Quantity 1 mL
Formula Weight 154.14 g/mol
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.