missing translation for 'onlineSavingsMsg'
Learn More

Terpineol, mixed isomers, 98%

Product Code. 15466618
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466618 2.5 L 2.5 L
15456618 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466618 Supplier Thermo Scientific Alfa Aesar Supplier No. H60146.K4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8000-41-7 | C10H18O | 154.25 g/mol

Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8000-41-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001557,MFCD00166983
InChI Key WUOACPNHFRMFPN-UHFFFAOYNA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

Melting Point 28°C to 36°C
Density 0.934
Boiling Point 213°C to 218°C
Flash Point 91°C (196°F)
Refractive Index 1.484
Quantity 2.5 L
Solubility Information Miscible with organic solvents. Slightly miscible with water.
Formula Weight 154.25
Percent Purity 98%
Chemical Name or Material Terpineol, mixed isomers

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.