missing translation for 'onlineSavingsMsg'
Learn More

gamma-Terpinene, 97%, stabilized

Product Code. p-3758770 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10052650

Brand: Thermo Scientific Acros 207501000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-85-4 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-85-4, 10191-41-0
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001537
InChI Key YKFLAYDHMOASIY-UHFFFAOYSA-N
Synonym gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen
PubChem CID 7461
ChEBI CHEBI:10577
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,4-diene
SMILES CC1=CCC(=CC1)C(C)C

Specifications

Color White
Density 0.8470g/mL
Boiling Point 182°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.477
Quantity 100 mL
Beilstein 05, 128
Merck Index 14, 9170
Specific Gravity 0.847
Solubility Information Solubility in water: less than 1%. Other solubilities: miscible with alcohol,ether
Stabilizer 0.1 % max. dl-alpha tocopherol
Formula Weight 136.24
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material γ-Terpinene, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • y-Terpinene, stabilized
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.