missing translation for 'onlineSavingsMsg'
Learn More

Terizidone, TRC

Product Code. 30007536
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007536 25 mg 25mg
30004879 1 g 1g
30005810 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007536 Supplier Toronto Research Chemicals Supplier No. TRCT11550025MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25683-71-0
Molecular Formula C14H14N4O4
Molecular Weight (g/mol) 302.29
Synonym 3-Isoxazolidinone, 4,4'-[1,4-phenylenebis(methylidynenitrilo)]bis-, 3-Isoxazolidinone, 4,4'-[p-phenylenebis(methylidynenitrilo)]di- (7CI,8CI), 4,4'-[1,4-Phenylenebis(methylidynenitrilo)]bis[3-isoxazolidinone], B 2360, Terivalidin, Terizidon, Terizidone
IUPAC Name 4-[(E)-[4-[(E)-(3-oxo-1,2-oxazolidin-4-yl)iminomethyl]phenyl]methylideneamino]-1,2-oxazolidin-3-one
SMILES O=C1NOCC1\N=C\c2ccc(\C=N\C3CONC3=O)cc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H14N4O4/c19-13-11(7-21-17-13)15-5-9-1-2-10(4-3-9)6-16-12-8-22-18-14(12)20/h1-6,11-12H,7-8H2,(H,17,19)(H,18,20)/b15-5+,16-6+
Formula Weight 302.1015
Chemical Name or Material Terizidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.