missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Teriparatide

Product Code. 16401798
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401798 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401798 Supplier MedChemExpress Supplier No. HYP0059
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Teriparatide (Human parathyroid hormone-(1-34)) is a PHT agonist. Teriparatide (Human parathyroid hormone-(1-34)) can be used for osteoporosis research.

Specifications

Purity 96.42%
CAS 52232-67-4
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 4117.72 g/mol
Product Type Reference compound
Quantity 5 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (24.29 mM∣Need ultrasonic)
Synonym Human parathyroid hormone-(1-34)∣ hPTH (1-34)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.