missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tergitol™ 15-S-9

Product Code. 16368237 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.50 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16368237 2.5 L 2.50 L
16388237 250 mL 250mL
16358237 1 L 1 L
16378237 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16368237 Supplier Thermo Scientific Chemicals Supplier No. 464250025
  • ONLINE EXCLUSIVE PRICE
    £152.00
    £136.65 / 2.5 L
    Save £15.35
    10% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68131-40-8 | [C2H4O]n C13H27O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-40-8
Molecular Formula [C2H4O]n C13H27O
MDL Number MFCD00132411
SMILES CCCCCCCC(CCCCC)OCCO-*

Specifications

Density 1.0100 g/mL
Quantity 2.5 L
Flash Point 193°C
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Tergitol* 15-S-9
compliance-icons compliance-icons
Product Identifier
  • Tergitol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Alcohols, C11-15, Polyethylene glycol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.