missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TergitolTM 15-S-40, 70% solution in water

Product Code. 16318247 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.50 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16318247 250 mL 250mL
16398237 1 L 1 L
16308247 2.5 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16318247 Supplier Thermo Scientific Chemicals Supplier No. 464262500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 68131-40-8 | [C2H4O]n C13H27O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-40-8
Molecular Formula [C2H4O]n C13H27O
MDL Number MFCD00132411
SMILES CCCCCCCC(CCCCC)OCCO-*

Specifications

Density 1.0720 g/mL
Quantity 250 mL
Specific Gravity 1.072
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Tergitol* 15-S-40

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.