missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Terfenadine

Product Code. 18095002
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095002 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18095002 Supplier R&D Systems™ Supplier No. 3948/50
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. H1 antagonist. Also KV11.1 (hERG) and Kir6 (KATP) channel blocker

Histamine H1 receptor antagonist. Also blocks KV11.1 (hERG) and Kir6 (KATP) channels (IC50 values are 204 nM and 1.2 μM respectively). Inhibits the delayed rectifier K+ current (IKr) in guinea pig ventricular myocytes (IC50 = 50 nM). Activity prolongs QT and induces Torsades de pointes (TdP); cardiotoxic in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50679-08-8
Quantity 50 mg
Product Line Tocris
Molecular Formula C32H41NO2
Formula Weight 471.67
Purity >98%
Molecular Weight (g/mol) 471.69
Chemical Name or Material Terfenadine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.