missing translation for 'onlineSavingsMsg'
Learn More

Terfenadine, MedChemExpress

Product Code. 16408676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408676 Supplier Medchem Express Supplier No. HYB1193
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Terfenadine ((±)-Terfenadine) is a potent open-channel blocker of hERG with an IC50 of 204 nM. Terfenadine, an H1 histamine receptor antagonist, acts as a potent apoptosis inducer in melanoma cells through modulation of Ca2+ homeostasis. Terfenadine induces ROS-dependent apoptosis, simultaneously activates Caspase-4, -2, -9.

Terfenadine ((±)-Terfenadine) is a potent open-channel blocker of hERG with an IC50 of 204 nM. Terfenadine, an H1 histamine receptor antagonist, acts as a potent apoptosis inducer in melanoma cells through modulation of Ca2+ homeostasis. Terfenadine induces ROS-dependent apoptosis, simultaneously activates Caspase-4, -2, -9.

Specifications

Chemical Name or Material Terfenadine
CAS 50679-08-8
Color White
Purity Grade Notes Research
Molecular Formula C32H41NO2
Quantity 100 mg
Synonym (±)-Terfenadine MDL-991
Solubility Information DMSO : ≥ 50 mg/mL (106.01 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(C1=CC=C(C(C)(C)C)C=C1)CCCN2CCC(C(C3=CC=CC=C3)(O)C4=CC=CC=C4)CC2
Molecular Weight (g/mol) 471.67
Formula Weight 471.67
Percent Purity 99.86%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.