Learn More
TERF2 Mouse anti-Human, Unconjugated, Clone: 5B1E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66893-1-IG-150UL
Description
The TERF2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TERF2. This antibody recognizes human, mouse, rat antigen. The TERF2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TERF2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 5B1E1 | |
| Unconjugated | |
| 150 μL | |
| 7014 | |
| IgG1 |