missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TERF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11899-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.Specifications
| TERF1 | |
| Polyclonal | |
| Unconjugated | |
| TERF1 | |
| hTRF1 AS, NIMA interacting protein 2, PIN2, t TRF1, Telomeric protein Pin2/TRF1, TERF1, TRBF1, TRF, TRF1, TTAGGG repeat binding factor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7013 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P54274 | |
| TERF1 | |
| TERF1 Fusion Protein Ag2491 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction