Learn More
TERF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11899-1-AP-150UL
 View more versions of this product
Product Code. 16869273
  • £349.00 / 150µL
Description
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
Specifications
Specifications
TERF1
Polyclonal
Unconjugated
TERF1
hTRF1 AS, NIMA interacting protein 2, PIN2, t TRF1, Telomeric protein Pin2/TRF1, TERF1, TRBF1, TRF, TRF1, TTAGGG repeat binding factor 1
Rabbit
Antigen Affinity Chromatography
RUO
7013
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P54274
TERF1
TERF1 Fusion Protein Ag2491
150 μL