missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic Acid, TRC

Product Code. 30003559
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003559 Supplier Toronto Research Chemicals Supplier No. TRCT1125005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-21-0
Molecular Formula C8 H6 O4
Quantity 5 g
Synonym Terephthalic Acid, 1,4-Benzenedicarboxylic acid, Terephthalic acid (7CI,8CI), 1,4-Dicarboxybenzene, 4-Carboxybenzoic acid, Amoco TA 33, NSC 36973, QTA, S-LOP, TA 33LP, TPA, TPA (terephthalic acid), WR 16262, p-Benzenedicarboxylic acid, p-Carboxybenzoic acid, p-Dicarboxybenzene, p-Phthalic acid
InChI Formula InChI=1S/C8H6O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h1-4H,(H,9,10)(H,11,12)
SMILES OC(=O)c1ccc(cc1)C(=O)O
IUPAC Name terephthalic acid
Molecular Weight (g/mol) 166.13
Formula Weight 166.0266
Chemical Name or Material Terephthalic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.