missing translation for 'onlineSavingsMsg'
Learn More

Terephthalic acid, MedChemExpress

Product Code. 16432678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432678 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16432678 Supplier Medchem Express Supplier No. HYW010098
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Terephthalic acid is one isomer of the three phthalic, a precursor to the polyester PET, used to make clothing and plastic bottles.

Terephthalic acid is one isomer of the three phthalic, a precursor to the polyester PET, used to make clothing and plastic bottles.

Specifications

Chemical Name or Material Terephthalic acid
CAS 100-21-0
Color White
Purity Grade Notes Research
Molecular Formula C8H6O4
Quantity 5 g
Solubility Information DMSO : 20 mg/mL (120.39 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(C(O)=O)C=C1
Molecular Weight (g/mol) 166.13
Formula Weight 166.13
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.