missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terephthalic acid, bis-ethyl ester 4000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Terephthalic Acid Diethyl Ester |
| CAS | 636-09-9 |
| Molecular Formula | C12 H14 O4 |
| Quantity | 1 mL |
| Synonym | 1,4-Benzenedicarboxylic acid, 1,4-diethyl ester, 1,4-Benzenedicarboxylic acid, diethyl ester (9CI), Terephthalic acid, diethyl ester (6CI,7CI,8CI), Diethyl terephthalate, NSC 68816, p-Benzenedicarboxylic acid diethyl ester, p-Diethyl phthalate, Terephthalic acid diethyl ester, Terephthalic acid, bis-ethyl ester |
| InChI Formula | InChI=1S/C12H14O4/c1-3-15-11(13)9-5-7-10(8-6-9)12(14)16-4-2/h5-8H,3-4H2,1-2H3 |
| SMILES | CCOC(=O)c1ccc(cc1)C(=O)OCC |
| IUPAC Name | diethyl benzene-1,4-dicarboxylate |
| Molecular Weight (g/mol) | 222.2372 |
| Formula Weight | 222.0892 g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction