missing translation for 'onlineSavingsMsg'
Learn More

Terephthaldicarboxaldehyde, 98%

Product Code. 10011610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011610 500 g Plastic bottle 500g
10153520 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011610 Supplier Thermo Scientific Acros Supplier No. 137895000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-27-8 | C8H6O2 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-27-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00006949
InChI Key KUCOHFSKRZZVRO-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
PubChem CID 12173
IUPAC Name terephthalaldehyde
SMILES C1=CC(=CC=C1C=O)C=O

Specifications

Melting Point 114°C to 116°C
Color White to Yellow
Boiling Point 245°C to 248°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H4(CHO)2
Quantity 500 g
Beilstein 07, 675
Solubility Information Solubility in water: 3g/L (50°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 134.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Terephthaldicarboxaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Terephthaldicarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.