missing translation for 'onlineSavingsMsg'
Learn More

Terephthaldehyde monodiethylacetal, 97%, stabilized

Product Code. 10207660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10207660 100 g Glass bottle 100g
10159562 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10207660 Supplier Thermo Scientific Acros Supplier No. 294631000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81172-89-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00010217
InChI Key HTMXMFARWHNJDW-UHFFFAOYSA-N
Synonym 4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
PubChem CID 595993
IUPAC Name 4-(diethoxymethyl)benzaldehyde
SMILES CCOC(OCC)C1=CC=C(C=O)C=C1

Specifications

Color Brown to White
Density 1.0400g/mL
Boiling Point 89°C to 93°C (7.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CHOC6H4CH(OC2H5)2
Refractive Index 1.5050 to 1.5075
Quantity 100 g
Specific Gravity 1.04
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,chlorinated solvents and,toluene
Stabilizer 0.1 % triethylamine
Formula Weight 208.26
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material Terephthaldehyde monodiethylacetal, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Terephthaldehyde monodiethylacetal, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.