missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehyde mono(diethyl acetal), 97%, stab.

Product Code. 11403960
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403960 5 g 5g
11418396 25 g 25g
11428396 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403960 Supplier Thermo Scientific Alfa Aesar Supplier No. A15413.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 81172-89-6 | C12H16O3 | 208.26 g/mol

4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Diethoxymethyl)benzaldehyde was used in the synthesis of 4-(diethoxymethyl)benzyl alcohol.

Solubility
soluble in alcohol, chlorinated solvents and toluene. Decomposes in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81172-89-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00010217
InChI Key HTMXMFARWHNJDW-UHFFFAOYSA-N
Synonym 4-diethoxymethyl benzaldehyde, terephthalaldehyde mono diethyl acetal, benzaldehyde, 4-diethoxymethyl, terephthaldehyde mono diethyl acetal, terephthalaldehyde mono-diethyl acetal, pubchem22621, acmc-209tbs, 4-diethoxymethyl-benzaldehyde, 4-diethoxymethy benzaldehyde, 4-diethyoxymethyl benzaldehyde
PubChem CID 595993
IUPAC Name 4-(diethoxymethyl)benzaldehyde
SMILES CCOC(OCC)C1=CC=C(C=O)C=C1

Specifications

Density 1.047
Boiling Point 89°C to 93°C (7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 5 g
Beilstein 4293872
Sensitivity Air Sensitive
Solubility Information soluble in alcohol,chlorinated solvents and toluene. Decomposes in water.
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material Terephthalaldehyde mono(diethyl acetal), stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.