missing translation for 'onlineSavingsMsg'
Learn More

Terephthalaldehyde, 98%

Product Code. 11479130
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11479130

Brand: Thermo Scientific Alfa Aesar A14930.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-27-8 | C8H6O2 | 134.134 g/mol

Terephthalaldehyde is used as an intermediate for the preparation of dyes and fluorescent whitening agents. It is used in the synthesis of polyimines by reacting with aliphatic diamines. It is used as a precursor for the preparation of paramagnetic microporous polymeric organic frameworks (POFs) through copolymerization with pyrrole, indole, and carbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Terephthalaldehyde is used as an intermediate for the preparation of dyes and fluorescent whitening agents. It is used in the synthesis of polyimines by reacting with aliphatic diamines. It is used as a precursor for the preparation of paramagnetic microporous polymeric organic frameworks (POFs) through copolymerization with pyrrole, indole, and carbazole.

Solubility
Soluble in hot water, alcohol and ether.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-27-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.134
MDL Number MFCD00006949
InChI Key KUCOHFSKRZZVRO-UHFFFAOYSA-N
Synonym 1,4-benzenedicarboxaldehyde, p-phthalaldehyde, terephthaldehyde, terephthaldialdehyde, terephthaladehyde, terephthaldicarboxaldehyde, terephthalic aldehyde, 1,4-phthalaldehyde, p-formylbenzaldehyde, terephtaldehyde
PubChem CID 12173
IUPAC Name terephthalaldehyde
SMILES C1=CC(=CC=C1C=O)C=O

Specifications

Melting Point 114°C to 117°C
Density 1.06
Boiling Point 245°C
Flash Point 76°C (168°F)
Quantity 500 g
Beilstein 385863
Sensitivity Air Sensitive
Solubility Information Soluble in hot water,alcohol and ether.
Formula Weight 134.14
Percent Purity ≥98%
Chemical Name or Material Terephthalaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.