missing translation for 'onlineSavingsMsg'
Learn More

Terconazole, MedChemExpress

Product Code. 16464627
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16464627

Brand: Medchem Express HYB1790

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terconazole is a broad-spectrum antifungal medication for the treatment of vaginal yeast infection.

Terconazole is a broad-spectrum antifungal medication for the treatment of vaginal yeast infection.

Specifications

Chemical Name or Material Terconazole
Physical Form Powder
CAS 67915-31-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H31Cl2N5O3
For Use With (Application) COVID-19-immunoregulation
Synonym R42470
Solubility Information DMSO : ≥ 30 mg/mL (56.34 mM)
SMILES CC(N1CCN(C2=CC=C(OC[C@@H]3O[C@@](CN4N=CN=C4)(C5=CC=C(Cl)C=C5Cl)OC3)C=C2)CC1)C
Molecular Weight (g/mol) 532.46
Formula Weight 532.46
Percent Purity 99.16%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.