missing translation for 'onlineSavingsMsg'
Learn More

Terbufoxon Sulfoxide, TRC

Product Code. 30138543
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138543 2.5 mg 2.50mg
30135108 10 mg 10mg
30008269 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138543 Supplier Toronto Research Chemicals Supplier No. TRCT1090852.5MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56165-57-2
Molecular Formula C9 H21 O4 P S2
Molecular Weight (g/mol) 288.36
Synonym Phosphorothioic acid, S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester, O,O-Diethyl S-(tert-butylsulfinyl)methyl phosphorothioate, Terbufos oxon sulfoxide, Terbufoxon sulfoxide
IUPAC Name 2-(diethoxyphosphorylsulfanylmethylsulfinyl)-2-methylpropane
SMILES CCOP(=O)(OCC)SCS(=O)C(C)(C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H21O4PS2/c1-6-12-14(10,13-7-2)15-8-16(11)9(3,4)5/h6-8H2,1-5H3
Formula Weight 288.0619
Chemical Name or Material Terbufos-oxon-sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.