missing translation for 'onlineSavingsMsg'
Learn More

Terbufos-sulfoxide, TRC

Product Code. 30410719
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410719 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410719 Supplier Toronto Research Chemicals Supplier No. TRCT10900325MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terbufos-sulfoxide
CAS 10548-10-4
Purity Grade Notes HPLC
Molecular Formula C9 H21 O3 P S3
Quantity 25 mg
Synonym Phosphorodithioic acid','S-[[(1,1-dimethylethyl)sulfinyl]methyl] O,O-diethyl ester (9CI','ACI), Methanethiol','(tert-butylsulfinyl)-','S-ester with O,O-diethyl phosphorodithioate (8CI), Phosphorodithioic acid','S-[(tert-butylsulfinyl)methyl] O,O-diethyl ester (7CI','8CI), Terbufos sulfoxide
InChI Formula InChI=1S/C9H21O3PS3/c1-6-11-13(14,12-7-2)15-8-16(10)9(3,4)5/h6-8H2,1-5H3
SMILES CCOP(=S)(OCC)SCS(=O)C(C)(C)C
IUPAC Name tert-butylsulfinylmethylsulfanyl-diethoxy-sulfanylidene-λ5-phosphane
Molecular Weight (g/mol) 304.43
Formula Weight 304.039
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.