missing translation for 'onlineSavingsMsg'
Learn More

Terbufos-d10 (O,O-diethyl-d10), TRC

Product Code. 30420420
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420420 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420420 Supplier Toronto Research Chemicals Supplier No. TRCT1090010.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terbufos-d10 (O,O-diethyl-d10)
CAS 1276197-42-2
Molecular Formula C9 D10 H11 O2 P S3
Quantity 0.5 mg
Synonym S-[[(1,1-Dimethylethyl)thio]methyl] O,O-di(ethyl-1,1,2,2,2-d5) phosphorodithioate, S-[(tert-Butylthio)methyl]phosphorodithioic acid O,O-di(ethyl-d5) ester, Terbufos-d10, Terbufos-D10 (diethoxy-D10), Terbufos-d10 (O,O-diethyl-d10), tert-Butylsulfanylmethylsulfanyl-bis(1,1,2,2,2-pentadeuterioethoxy)-thioxo-λμ-phosphane
InChI Formula InChI=1S/C9H21O2PS3/c1-6-10-12(13,11-7-2)15-8-14-9(3,4)5/h6-8H2,1-5H3/i1D3,2D3,6D2,7D2
SMILES [2H]C([2H])([2H])C([2H])([2H])OP(=S)(OC([2H])([2H])C([2H])([2H])[2H])SCSC(C)(C)C
IUPAC Name tert-butylsulfanylmethylsulfanyl-bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-λ5-phosphane
Molecular Weight (g/mol) 298.492
Formula Weight 298.107
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.