missing translation for 'onlineSavingsMsg'
Learn More

Terbufos 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30323153
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323153 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323153 Supplier Dr. Ehrenstorfer Supplier No. DREL17270000AL
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terbufos
CAS 13071-79-9
Analyte or Component Names Terbufos
Shipping Condition Room Temperature
Molecular Formula C9 H21 O2 P S3
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphorodithioic acid, S-[(tert-butylthio)methyl] O,O-diethyl ester (6CI,7CI,8CI), Methanethiol, (tert-butylthio)-, S-ester with O,O-diethyl phosphorodithioate (8CI), AC 92100, Counter, Counter 15G, Cyanater, O,O-Diethyl S-(tert-butylthio)methyl phosphorodithioate, O,O-Diethyl S-[[(1,1-dimethylethyl)thio]methyl] phosphorodithioate, S-[(tert-Butylthio)methyl] O,O-diethyl phosphorodithioate, Terbufos, Terbuphos, Terbutifos
InChI Formula InChI=1S/C9H21O2PS3/c1-6-10-12(13,11-7-2)15-8-14-9(3,4)5/h6-8H2,1-5H3
Solution Type Single Solution
SMILES CCOP(=S)(OCC)SCSC(C)(C)C
IUPAC Name tert-butylsulfanylmethylsulfanyl-diethoxy-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 288.43
Quantity 10 mL
Formula Weight 288.0441 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.