missing translation for 'onlineSavingsMsg'
Learn More

Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

Catalog Number 11365108
Click to view available options
Quantity:
10 mm
25 mm
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]

Form: Approx. 1.04g/mm; Cast rod, diameter is approximate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Physical Form Cast Rod
Quantity 10 mm
Assay Percent Range 99.9% (Metals basis)
Appearance Silver Gray rod
Chemical Name or Material Terbium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'