missing translation for 'onlineSavingsMsg'
Learn More

Terbium plasma standard solution, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 013860.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11361429

  • £54.00 / 100mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Tellurium
Liquid
MFCD00011256
7732-18-5
Tb4O7 in 5% HNO3

Specifications

Tellurium
Liquid
Tb4O7 in 5% HNO3
Nitric acid: 5%; Terbium(III, IV) oxide: 0.12%; Water: 94.88%
100mL
7732-18-5
Liquid
MFCD00011256
23 hPa (17mm Hg) at 20°C
Videos
SDS