missing translation for 'onlineSavingsMsg'
Learn More

Terbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11361609
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]
This item is not returnable. View return policy
7440-27-9
Tb
158.93
MFCD00011256
GZCRRIHWUXGPOV-UHFFFAOYSA-N
unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
23958
CHEBI:33376
terbium
[Tb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver to Gray
UN Number UN3178
Quantity 1 g
Percent Purity 99.9%
Odor Odorless
Form Pieces
Assay (REO)
Chemical Name or Material Terbium pieces, Distilled dendritic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'