missing translation for 'onlineSavingsMsg'
Learn More

Terbium pieces, 99.9% (REO)

Product Code. p-4824980
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11326527

Brand: Thermo Scientific Alfa Aesar 000324.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Pieces
Quantity 1 g
Assay Percent Range 99.9% (REO)
Odor Odorless
Appearance Silver Gray pieces
Chemical Name or Material Terbium
compliance-icons
Product Identifier
  • Terbium pieces
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use to extinguish.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.