missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) trifluoromethanesulfonate, 98%

Product Code. 11343868
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343868 50 g 50g
11353868 2 g 2g
11333868 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343868 Supplier Thermo Scientific Alfa Aesar Supplier No. 040312.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 148980-31-8 | C3F9O9S3Tb | 606.115 g/mol

It is used as a reagent & chemical intermediates. It is a raw material used in synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148980-31-8
Molecular Formula C3F9O9S3Tb
Molecular Weight (g/mol) 606.115
MDL Number MFCD00192461
InChI Key JLYAVYSVLLIWEK-UHFFFAOYSA-K
Synonym terbium iii trifluoromethanesulfonate, terbium tritriflate, terbium iii triflate, terbium tris trifluoromethanesulfonate, trifluoromethanesulfonic acid terbium iii salt, terbium trifluoromethanesulfonate, terbium triflate, tb otf 3, acmc-1byi6
PubChem CID 11786785
IUPAC Name terbium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tb+3]

Specifications

Physical Form Powder
Quantity 50 g
Formula Weight 606.12
Percent Purity 98%
Chemical Name or Material Terbium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.