missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals

Product Code. 11420301
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420301 2 g 2g
11430301 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420301 Supplier Thermo Scientific Alfa Aesar Supplier No. 040573.NA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In observation of slow magnetic relaxation in triple-stranded lanthanide helicates Terbium(III) perchlorate is substituted for dysprosium(III) perchlorate hexahydrate. By reacting 3-formylchromone and hydrated terbium (III) perchlorate in isopropanol, 3-hydroxymethylchromone [icm], bis(3-chromonyl)methanol [dcm] and the novel complex Tb(imc)3(ClO4)3.5H2O are obtained.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-09-6
MDL Number MFCD00150362

Specifications

Color Colorless
Physical Form Liquid
Quantity 2 g
Linear Formula Tb(ClO4)3·6H2O
UN Number UN3211
Formula Weight 565.37 (457.28 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Density 1.5g/mL
Chemical Name or Material Terbium(III) perchlorate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.