missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III,IV) oxide, REacton™, 99.9% (REO)

Product Code. 11346248
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346248 1 kg 1kg
11336248 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346248 Supplier Thermo Scientific Alfa Aesar Supplier No. 011207.A1
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12037-01-3 | Tb{4}O{7}

Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 2340°C
CAS 12037-01-3
Physical Form Solid
Quantity 1 kg
Molecular Formula Tb{4}O{7}
Percent Purity 99.9%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.