missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III,IV) oxide, REacton™, 99.99% (REO)

Product Code. 11356248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356248 25 g 25g
11366248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356248 Supplier Thermo Scientific Alfa Aesar Supplier No. 011208.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12037-01-3 | O7Tb4 | 747.69 g/mol

Terbium(III,IV) oxide is used in the preparation of other terbium compounds. Activating agents for fluorescent powders, composites for catalytic automobile exhaust converters, Glass materials with a Faraday Rotation effect for optical and laser-based devices, Magneto-optical glasses, Magneto-optical recording materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-01-3
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

Specifications

Melting Point 2340°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (REO)
Solubility Information soluble in dilute acids. Insoluble in water.
Formula Weight 747.69
Chemical Name or Material Terbium(III, IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.