missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

Product Code. 11386908
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386908 25 g 25g
11396908 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386908 Supplier Thermo Scientific Alfa Aesar Supplier No. 013657.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13708-63-9 | F3Tb | 215.92 g/mol

Terbium(III) fluoride is used as a source of terbium. It is also useful in the preparation of fluoride glasses. Further, it is used in the preparation of electroluminescent thin films and luminescent zinc sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13708-63-9
Molecular Formula F3Tb
Molecular Weight (g/mol) 215.92
MDL Number MFCD00049572
InChI Key LKNRQYTYDPPUOX-UHFFFAOYSA-K
IUPAC Name terbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Tb+3]

Specifications

Melting Point 1172°C
Physical Form Crystalline
Quantity 25 g
UN Number UN3288
Solubility Information Slightly soluble in mineral acids. Insoluble in water.
Formula Weight 215.92
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.