missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Terbium(III) chloride hexahydrate is used in the primary chemical intermediates and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Merck Index | 14,9157 |
Solubility Information | soluble in water |
Formula Weight | 373.38 (265.29 Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Odor | Odorless |
Assay | (REO) |
Chemical Name or Material | Terbium(III) chloride hexahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'