missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Product Code. p-4408185
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11396248

Brand: Thermo Scientific Alfa Aesar 011209.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13798-24-8 | Cl3H12O6Tb | 373.37 g/mol

Terbium(III) chloride hexahydrate is a water soluble source used for the preparation of metallic terbium, which is used in electronic devices and an additive for special alloy namely Terfenol-D. It is used in the preparation of complexes, which are used in the spectrofluorimetric determination of various chemicals in pharmaceutical formulations. Further, it finds use in the preparation of terbium nanotubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13798-24-8
Molecular Formula Cl3H12O6Tb
Molecular Weight (g/mol) 373.37
MDL Number MFCD00149870
InChI Key ULJUVCOAZNLCJZ-UHFFFAOYSA-K
IUPAC Name terbium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Very soluble in water.
Formula Weight 373.38 (265.29 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Terbium(III) chloride hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.