missing translation for 'onlineSavingsMsg'
Learn More

Terbium(III) bromide, ultra dry, 99.99% (REO)

Product Code. 11368208
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368208 1 g 1g
11358208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368208 Supplier Thermo Scientific Alfa Aesar Supplier No. 039425.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14456-47-4 | Br3Tb | 398.637 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-47-4
Molecular Formula Br3Tb
Molecular Weight (g/mol) 398.637
MDL Number MFCD00049571
InChI Key AZNZWHYYEIQIOC-UHFFFAOYSA-K
Synonym terbium iii bromide, terbium tribromide, terbium 3+ tribromide, terbium iii ; bromide, acmc-1btzk, terbium 3+ ion tribromide, terbium bromide
PubChem CID 84452
IUPAC Name terbium(3+);tribromide
SMILES [Br-].[Br-].[Br-].[Tb+3]

Specifications

Boiling Point 1490°C
Melting Point 827°C
Physical Form Powder
Quantity 1 g
Formula Weight 398.65
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Terbium(III) bromide, Ultra dry

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.