Learn More
Terbium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
£100.00 - £335.00
Chemical Identifiers
CAS | 100587-92-6 |
---|---|
Molecular Formula | C6H9O6Tb |
Molecular Weight (g/mol) | 336.06 |
MDL Number | MFCD00149233 |
InChI Key | JQBILSNVGUAPMM-UHFFFAOYSA-K |
IUPAC Name | terbium(3+) triacetate |
SMILES | [Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11312558
|
Thermo Scientific Alfa Aesar
014600.09 |
10 g |
£100.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11302558
|
Thermo Scientific Alfa Aesar
014600.18 |
50 g |
£335.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Terbium Acetate has important role as an activator for green phosphors used in color TV tubes. Meanwhile Terbium Acetate is also used in special lasers and as a dopant in solid-state devices. Terbium is also used in alloys and in the production of electronic devices. As a component of Terfenol-D, Terbium is of use in actuators, in naval sonar systems, sensors, in the SoundBugdevice (its first commercial application), and other magnetomechanical devices. It is also used in alloys and in the production of electronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100587-92-6 | |
336.06 | |
JQBILSNVGUAPMM-UHFFFAOYSA-K | |
[Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
C6H9O6Tb | |
MFCD00149233 | |
terbium(3+) triacetate |
Specifications
100587-92-6 | |
99.9% (REO) | |
MFCD00149233 | |
JQBILSNVGUAPMM-UHFFFAOYSA-K | |
terbium(3+) triacetate | |
336.08 (anhydrous) | |
Terbium(III) acetate hydrate |
Crystalline Aggregates | |
C6H9O6Tb | |
Soluble in water,moderately soluble in strong mineral acids | |
[Tb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
336.06 | |
Hygroscopic |
Safety and Handling
missing translation for 'einecsNumber' : 247-065-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only