missing translation for 'onlineSavingsMsg'
Learn More

Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO)

Product Code. p-4824979
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11321799

Brand: Thermo Scientific Alfa Aesar 012430.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.52g/25 x 25mm | CAS: 7440-27-9 | Tb | 158.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-27-9
Molecular Formula Tb
Molecular Weight (g/mol) 158.93
MDL Number MFCD00011256
InChI Key GZCRRIHWUXGPOV-UHFFFAOYSA-N
Synonym unii-06ssf7p179, ingot, foil, terbio, atom, chips, pieces, powder, acmc-20ajly, pieces, distilled dendritic
PubChem CID 23958
ChEBI CHEBI:33376
IUPAC Name terbium
SMILES [Tb]

Specifications

Color Silver to Gray
Physical Form Foil
Quantity 25 x 25 mm
Assay Percent Range 99.9% (REO)
Appearance Silver Gray foil
Chemical Name or Material Terbium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.