missing translation for 'onlineSavingsMsg'
Learn More

Terbinafine hydrochloride, MedChemExpress

Product Code. 16401256
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
This item is not returnable. View return policy

Product Code. 16401256

Brand: Medchem Express HY17395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. Terbinafine hydrochloride is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Terbinafine hydrochloride (TDT 067 hydrochloride) is an orally active and potent antifungal agent. Terbinafine hydrochloride is a potent non-competitive inhibitor of squalene epoxidase from Candida, with a Ki of 30 nM. Terbinafine hydrochloride also shows antibacterial activity against certain Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Terbinafine hydrochloride
Physical Form Solid
CAS 78628-80-5
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C21H26ClN
For Use With (Application) Neuroscience-Neuromodulation
Synonym TDT 067 hydrochloride
Solubility Information DMSO : 60 mg/mL (182.99 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.05 mM; ultrasonic and warming and heat to 60°C)
SMILES CN(C/C([H])=C([H])/C#CC(C)(C)C)CC1=CC=CC2=CC=CC=C21.[H]Cl
Molecular Weight (g/mol) 327.89
Formula Weight 327.89
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.