missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terazosin Hydrochloride Dihydrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Terazosin hydrochloride dihydrate |
| CAS | 70024-40-7 |
| Analyte or Component Names | Terazosin hydrochloride dihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H25 N5 O4 . Cl H . 2 H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Terazosin Hydrochloride Dihydrate, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine Monohydrochloride Dihydrate, Terazosin Monohydrochloride Dihydrate |
| InChI Formula | InChI=1S/C19H25N5O4.ClH.2H2O/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;;;/h10-11,14H,3-9H2,1-2H3,(H2,20,21,22);1H;2*1H2 |
| Solution Type | Neat |
| SMILES | O.O.Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4CCCO4 |
| Show More |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion